Barium 2-ethylhexoxide in hexanes/toluene

Supplier: STREM CHEMICALS INC MS
Danger

56-4000-25G 56-4000-5G 56-4000-100G
100210-010EA 1933.42 USD
100210-010 100210-012 100198-856
Barium 2-ethylhexoxide in hexanes/toluene
Barium 2-ethylhexoxide

Alkoxide

Flash Pt: –9 °F
MDL Number: MFCD00079662
CAS Number: 29170-99-8
UN: 1992
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR