Cesium carborane

Supplier: STREM CHEMICALS INC MS
Warning

55-5525-250MG 55-5525-1G
100197-560EA 441.6 USD
100197-560 100207-410
Cesium carborane
Cesium carborane
MDL Number: MFCD01634611
CAS Number: 12539-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR