(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')cesium
Supplier: STREM CHEMICALS INC MS
Synonyms:
Cs(TMHD), 2,2,6,6-Tetramethyl-3,5-heptanedionato cesium
55-5000-1G
55-5000-5G
100201-704EA
62.28
USD
100201-704
100205-964
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')cesium
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')cesium
Metal TMHD
Formula:
C₁₁H₁₉CsO₂ MW: 316.173 g/mol |
MDL Number:
MFCD00269816 CAS Number: 61346-75-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...