Cesium dodecahydrododecaborate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

55-1800-100G 55-1800-5G 55-1800-25G
100201-238EA 2598.65 USD
100201-238 100198-222 100206-112
Cesium dodecahydrododecaborate ≥98%
Cesium dodecahydrododecaborate
MDL Number: MFCD00273353
CAS Number: 12008-75-2
UN: 3178
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR