Tris(dimethylamino)antimony ≥99.99% (Sb basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

51-5000-25G 51-5000-1G 51-5000-5G
100203-228EA 1597.97 USD
100203-228 100206-760 100208-448
Tris(dimethylamino)antimony ≥99.99% (Sb basis), PURATREM
Tris(dimethylamino)antimony

Metal Alkylamides, Volatile Precursors for CVD

Boiling Pt: 32…34 °C (0.45 mmHg)
Density: 1,325
MDL Number: MFCD00269812
CAS Number: 7289-92-1
UN: 3399
ADR: 4.3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR