Tin(II) ethoxide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

50-2200-2G 50-2200-10G 50-2200-50G
100208-934EA 89.17 USD
100208-934 100199-244 100199-246
Tin(II) ethoxide ≥99%
Tin (II) ethoxide

Alkoxide

Formula: C₄H₁₀O₂Sn
MW: 208.83 g/mol
Melting Pt: 200 °C (dec.)
MDL Number: MFCD00015653
CAS Number: 14791-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR