Tin(IV) tert-butanolate ≥99.99% (Sn basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tin(IV) tert-butoxide, Tin(IV) t-butoxide

50-2100-25G 50-2100-5G 50-2100-1G
100203-222EA 819.51 USD
100203-222 100203-224 100210-000
Tin(IV) tert-butanolate ≥99.99% (Sn basis), PURATREM
Tin (IV) tert-butanolate

Alkoxide, Metal Alkoxides, Volatile Precursors for CVD

Formula: C₁₆H₃₆O₄Sn
MW: 411.165 g/mol
Boiling Pt: 65 °C (0.3 mmHg)
Melting Pt: 45 °C
Density: 1.06
MDL Number: MFCD00156551
CAS Number: 36809-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR