Tin(II) acetylacetonate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
50-1977-50G
50-1977-10G
100209-998EA
1680.06
USD
100209-998
100209-996
Tin(II) acetylacetonate ≥98%
Tin (II) acetylacetonate
Metal Beta-diketonates, Volatile Precursors for CVD
Formula:
C₁₀H₁₄O₄Sn Boiling Pt: 110 °C (0.1 mmHg) Density: 1,45 |
MDL Number:
MFCD00078005 CAS Number: 16009-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...