Bis(trimethylstannyl)acetylene ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Bis(trimethyltin)acetylene

50-0900-5G 50-0900-1G
100200-282EA 390.65 USD
100200-282 100204-084
Bis(trimethylstannyl)acetylene ≥99%
Bis(trimethylstannyl)acetylene
Formula: C₈H₁₈Sn₂
Melting Pt: 56 °C
MDL Number: MFCD01073798
CAS Number: 2117-50-2
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR