Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')indium(III) ≥99% (99.9%-In)

Supplier: STREM CHEMICALS INC MS

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)indium(III), In(TMHD)3, Indium(2,2,6,6-tetramethyl-3,5-heptanedionate)

49-2200-1G 49-2200-5G
100206-744EA 142.95 USD
100206-744 100206-746
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')indium(III) ≥99% (99.9%-In)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')indium(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₃₃H₅₇InO₆
MW: 664.63 g/mol
Melting Pt: 167 °C
MDL Number: MFCD00236550
CAS Number: 34269-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR