Indium(III) trifluoromethanesulfonate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Indium(III) trifluoromethanesulphonate, Indium triflate
49-1800-5G
49-1800-25G
100197-552EA
106.15
USD
100197-552
100209-992
Indium(III) trifluoromethanesulfonate ≥99%
Indium(III) trifluoromethanesulphonate
Metal Triflate
Formula:
C₃F₉InO₉S₃ MW: 562.03 g/mol Melting Pt: Dec. |
MDL Number:
MFCD00144478 CAS Number: 128008-30-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...