Trimethylphosphine(hexafluoroacetylacetonato)silver(I) ≥99% (99.9%-Ag)

Supplier: STREM CHEMICALS INC MS
Warning

47-3000-25G 47-3000-5G
100201-234EA 1496.06 USD
100201-234 100203-216
Trimethylphosphine(hexafluoroacetylacetonato)silver(I) ≥99% (99.9%-Ag)
Trimethylphosphine(hexafluoroacetylacetonato)silver(I)

Metal Beta-diketonates, Volatile Precursors for CVD

Boiling Pt: 95 °C (0.1 mmHg) (subl.)
Melting Pt: 140…142 °C
MDL Number: MFCD00269843
CAS Number: 148630-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR