(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')silver(I) ≥99.9% (Ag basis)
Supplier: STREM CHEMICALS INC MS
47-2600-25G
47-2600-1G
47-2600-5G
100203-214EA
1524.37
USD
100203-214
100206-736
100209-364
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')silver(I) ≥99.9% (Ag basis)
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')silver(I)
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formula:
C₁₁H₁₉AgO₂ Melting Pt: 178 °C (dec.) |
MDL Number:
MFCD01631284 CAS Number: 79827-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...