Silver neodecanoate ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

47-1050-5G 47-1050-1G
101442-470EA 288.74 USD
101442-470 101442-468
Silver neodecanoate ≥97%
Silver neodecanoate
Formula: C₁₀H₁₉AgO₂
MW: 279.120 g/mol
MDL Number: MFCD01940647
CAS Number: 68683-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR