Tetrakis(acetonitrile)palladium(II) tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

46-2145-1G 46-2145-250MG 46-2145-5G
100207-404EA 176.92 USD
100207-404 100206-726 100196-956
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate ≥98%
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate
Formula: C₈H₁₂B₂F₈N₄Pd
MDL Number: MFCD00043297
CAS Number: 21797-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR