Tetraamminepalladium(II) nitrate solution (5.0 % Pd) (w/w)

Supplier: STREM CHEMICALS INC MS

46-2140-50G 46-2140-10G
100196-288EA 1251.2 USD
100196-288 100200-476
Tetraamminepalladium(II) nitrate solution (5.0 % Pd) (w/w)
Tetraamminepalladium(II) nitrate
Formula: H₁₂N₆O₆Pd
MW: 298.552 g/mol
MDL Number: MFCD00011591
CAS Number: 13601-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR