Palladium(II) oxide ≥99.9% (Pd basis)

Supplier: STREM CHEMICALS INC MS

46-2110-5G 46-2110-1G
100209-362EA 1217.23 USD
100209-362 100209-360
Palladium(II) oxide ≥99.9% (Pd basis)
Palladium(II) oxide
Formula: OPd
Melting Pt: 870 °C
Density: 8.70
MDL Number: MFCD00011172
CAS Number: 1314-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR