Palladium 5% on calcium carbonate, dry, unreduced (Escat™ 1371)
Supplier: STREM CHEMICALS INC MS
46-2022-25G
46-2022-5G
100196-282EA
1167.69
USD
100196-282
100196-284
Palladium 5% on calcium carbonate, dry, unreduced (Escat™ 1371)
Palladium
Escat
Formula:
Pd MW: 106.420 g/mol |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...