Palladium 5% on alumina, dried powder, reduced (Escat™ 1241)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
46-1951-5G
46-1951-25G
100206-720EA
90.58
USD
100206-720
100206-718
Palladium 5% on alumina, dried powder, reduced (Escat™ 1241)
Palladium
Escat
Formula:
Pd MW: 106.420 g/mol |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...