Palladium 5% on alumina, dried powder, reduced

Supplier: STREM CHEMICALS INC MS
Warning

46-1950-5G 46-1950-25G 46-1950-100G
100197-538EA 114.65 USD
100197-538 100203-792 100197-536
Palladium 5% on alumina, dried powder, reduced
Palladium
Formula: Pd
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR