Palladium 0.5% on alumina, reduced
Supplier: STREM CHEMICALS INC MS
|
Warning
|
46-1920-25G
46-1920-100G
100207-696EA
302.89
USD
100207-696
100196-274
Palladium 0.5% on alumina, reduced
Palladium
Formula:
Pd |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...