Palladium 5% on activated carbon wetted with ca. 50% water, paste, reduced (Escat™ 1961)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
46-1909-10G
46-1909-50G
101419-088EA
615.69
USD
101419-088
101419-090
Palladium 5% on activated carbon wetted with ca. 50% water, paste, reduced (Escat™ 1961)
Palladium
Escat
Formula:
Pd MW: 106.420 g/mol |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...