Palladium 5% on activated peat carbon wetted with ca. 50% water, paste, reduced (Escat™ 1621)

Supplier: STREM CHEMICALS INC MS
Warning

46-1901-10G 46-1901-50G
100198-832EA 215.14 USD
100198-832 100198-834
Palladium 5% on activated peat carbon wetted with ca. 50% water, paste, reduced (Escat™ 1621)
Palladium

Escat

Formula: Pd
MW: 106.420 g/mol
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR