Palladium 10% on activated carbon, dried powder, reduced
Supplier: STREM CHEMICALS INC MS
|
Danger
|
46-1900-50G
46-1900-10G
100200-702EA
1406.89
USD
100200-702
100200-700
Palladium 10% on activated carbon, dried powder, reduced
Palladium
Formula:
Pd |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 UN: 3178 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...