Palladium 20% on activated carbon wetted with ca. 50% water, paste, unreduced (Pearlman's catalyst) (Escat™ 1951)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
46-1707-25G
46-1707-5G
100203-200EA
1108.25
USD
100203-200
100203-202
Palladium 20% on activated carbon wetted with ca. 50% water, paste, unreduced (Pearlman's catalyst) (Escat™ 1951)
Palladium
Escat
Formula:
Pd |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...