Palladium 5% on activated carbon wetted with 50-70% water, powder Evonik Noblyst® P1086
Supplier: STREM CHEMICALS INC MS
|
Warning
|
46-1703-50G
46-1703-10G
100205-142EA
893.11
USD
100205-142
100205-140
Palladium 5% on activated carbon wetted with 50-70% water, powder Evonik Noblyst® P1086
Palladium
Formula:
Pd MW: 106.420 g/mol |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...