[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: [1,2-Bis(diphenylphosphino)ethane]dichloropalladium(II), Dichloro[1,2-bis(diphenylphosphino)ethane]palladium(II)

46-0800-1G 46-0800-5G
100208-918EA 144.37 USD
100208-918 100208-920
[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride ≥98%
[1,2-Bis(diphenylphosphino)ethane]palladium(II) dichloride
Formula: C₂₆H₂₄Cl₂P₂Pd
Melting Pt: > 300 °C
MDL Number: MFCD00015702
CAS Number: 19978-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR