Dichloro(1,5-cyclooctadiene)palladium(II) ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
1,5-Cyclooctadienepalladium(II) Dichloride
46-0650-1G
46-0650-5G
100201-672EA
141.54
USD
100201-672
100201-674
Dichloro(1,5-cyclooctadiene)palladium(II) ≥99%
Dichloro(1,5-cyclooctadiene)palladium(II)
Formula:
C₈H₁₂Cl₂Pd MW: 285.50 g/mol Melting Pt: 205 °C (dec.) |
MDL Number:
MFCD00012412 CAS Number: 12107-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...