trans-Bis(triphenylphosphine)palladium(II) chloride ≥99% (99.9+%-Pd)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: trans-Dichlorobis(triphenylphosphine)palladium(II)

46-0530-1G 46-0530-5G
100209-976EA 83.51 USD
100209-976 100209-978
trans-Bis(triphenylphosphine)palladium(II) chloride ≥99% (99.9+%-Pd)
trans-Bis(triphenylphosphine)palladium(II) chloride
Formula: C₃₆H₃₀Cl₂P₂Pd
MW: 701.91 g/mol
MDL Number: MFCD00009593
CAS Number: 13965-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR