[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: [1,1'-Bis(di-tert-butylphosphino)ferrocene]dichloropalladium(II), 1,1'-Bis(di-tert-butylphosphino)ferrocene-palladium dichloride, Dichloro[1,1'-bis(di-t-butylphosphino)ferrocene]palladium(II), Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)

46-0445-2G 46-0445-500MG
101442-346EA 668.06 USD
101442-346 101442-344
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride ≥99%
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride
Formula: C₂₆H₄₄Cl₂FeP₂Pd
MW: 651.75 g/mol
MDL Number: MFCD08064219
CAS Number: 95408-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR