Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Palladium(I) tri-tert-butylphosphine bromide dimer, Di-mu-bromobis(tri-t-butylphosphino)dipalladium(I)

46-0355-100MG 46-0355-500MG
100208-440EA 92 USD
100208-440 100208-442
Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I)
Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I)
Formula: C₂₄H₅₆Br₂P₂Pd₂
MW: 779.3 g/mol
MDL Number: MFCD04114019
CAS Number: 185812-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR