Bis(triphenylphosphine)palladium(II) diacetate ≥99%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Bis(triphenylphosphine)palladium(II) acetate, Bis(acetato)bis(triphenylphosphine)palladium(II), Diacetatobis(triphenylphosphine)palladium(II)
46-0300-1G
46-0300-5G
100199-942EA
168.43
USD
100199-942
100199-944
Bis(triphenylphosphine)palladium(II) diacetate ≥99%
Bis(triphenylphosphine)palladium(II) diacetate
Formula:
C₄₀H₃₆O₄P₂Pd MW: 749.09 g/mol |
MDL Number:
MFCD00010013 CAS Number: 14588-08-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...