trans-Di(mu-acetato)bis[o-(di-o-tolylphosphino)benzyl]dipalladium(II) ≥97%
Supplier: STREM CHEMICALS INC MS
Synonyms:
trans-Di-μ-acetatobis[2-[bis(2-methylphenyl)phosphino]benzyl]dipalladium, cataCXium® C
46-0290-1G
46-0290-5G
100206-708EA
137.29
USD
100206-708
100206-710
trans-Di(mu-acetato)bis[o-(di-o-tolylphosphino)benzyl]dipalladium(II) ≥97%
trans-Di(mu-acetato)bis[o-(di-o-tolylphosphino)benzyl]dipalladium(II)
cataCXium
Formula:
C₄₆H₄₆O₄P₂Pd₂ MW: 937.6400 g/mol |
MDL Number:
MFCD01075746 CAS Number: 172418-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...