(1-Methylallyl)palladium chloride dimer ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

46-0275-2G 46-0275-500MG
100200-988EA 475.57 USD
100200-988 100200-986
(1-Methylallyl)palladium chloride dimer ≥99%
(1-Methylallyl)palladium chloride dimer
Formula: C₈H₁₄Cl₂Pd₂
MDL Number: MFCD01074457
CAS Number: 12081-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR