1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene(1,4-naphthoquinone)palladium(0) dimer ≥96%

Supplier: STREM CHEMICALS INC MS

46-0265-1G 46-0265-250MG
100198-818EA 324.12 USD
100198-818 100209-968
1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene(1,4-naphthoquinone)palladium(0) dimer ≥96%
1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene(1,4-naphthoquinone)palladium(0) dimer

NHC

MDL Number: MFCD08705778
CAS Number: 467220-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR