Bis(tricyclohexylphosphine)palladium(0) ≥98%

Supplier: STREM CHEMICALS INC MS

46-0260-250MG 46-0260-1G
100196-266EA 135.88 USD
100196-266 100206-706
Bis(tricyclohexylphosphine)palladium(0) ≥98%
Bis(tricyclohexylphosphine)palladium(0)
Formula: C₃₆H₆₆P₂Pd
MDL Number: MFCD01073796
CAS Number: 33309-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR