Bis(tri-tert-butylphosphine)palladium(0) ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Bis(tri-tert-butylphosphine)palladium, Bis(tri-t-butylphosphine)palladium(0)

46-0252-250MG 46-0252-1G
100196-264EA 150.03 USD
100196-264 100206-704
Bis(tri-tert-butylphosphine)palladium(0) ≥98%
Bis(tri-tert-butylphosphine)palladium(0)
Formula: C₂₄H₅₄P₂Pd
MW: 511.05 g/mol
MDL Number: MFCD03094580
CAS Number: 53199-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR