Tetrakis[(R)-(+)-N-(p-dodecylphenylsulfonyl)prolinato]dirhodium(II)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tetrakis[(R)-N-(p-dodecylphenylsulphonyl)prolinato]dirhodium(II), Rh2(R-DOSP)4

45-2100-50MG 45-2100-250MG
100211-112EA 66.52 USD
100211-112 100211-114
Tetrakis[(R)-(+)-N-(p-dodecylphenylsulfonyl)prolinato]dirhodium(II)
Tetrakis[(R)-(+)-N-(p-dodecylphenylsulfonyl)prolinato]dirhodium(II)
Formula: C₉₂H₁₄₄N₄O₁₆Rh₂S₄
MW: 1896.22 g/mol
MDL Number: MFCD04974245
CAS Number: 178879-60-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR