Rhodium(II) trifluoroacetate dimer ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

45-1960-2G 45-1960-500MG
100197-530EA 1102.58 USD
100197-530 100199-938
Rhodium(II) trifluoroacetate dimer ≥95%
Rhodium(II) trifluoroacetate dimer
Formula: C₈F₁₂O₈Rh₂
MDL Number: MFCD00209611
CAS Number: 31126-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR