Rhodium 5% on activated wood carbon wetted with ca. 50% water, paste, reduced (Escat™ 3401)

Supplier: STREM CHEMICALS INC MS
Warning

45-1875-5G 45-1875-1G
100207-694EA 747.32 USD
100207-694 100207-692
Rhodium 5% on activated wood carbon wetted with ca. 50% water, paste, reduced (Escat™ 3401)
Rhodium

Escat

Formula: Rh
MW: 102.9 g/mol
MDL Number: MFCD00011201
CAS Number: 7440-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR