Rhodium 5% on alumina
Supplier: STREM CHEMICALS INC MS
|
Warning
|
45-1830-5G
45-1830-1G
45-1830-25G
100210-816EA
1184.68
USD
100210-816
100198-206
100200-472
Rhodium 5% on alumina
Rhodium
Formula:
Rh |
MDL Number:
MFCD00011201 CAS Number: 7440-16-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...