Rhodium(II) octanoate dimer ≥98%
Supplier: STREM CHEMICALS INC MS
45-1809-5G
45-1809-500MG
45-1809-1G
100199-416EA
1941.91
USD
100199-416
100196-256
100207-690
Rhodium(II) octanoate dimer ≥98%
Rhodium(II) octanoate dimer
Formula:
C₃₂H₆₀O₈Rh₂ MW: 778.63 g/mol |
MDL Number:
MFCD00064724 CAS Number: 73482-96-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...