Rhodium (III) acetylacetonate ≥97% (99.9%-Rh)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tris(2,4-pentanedionato-O,O')rhodium (III), Rhodium (III) tris(2,4-pentanedionato-O,O'), Rhodium (III) 2,4-pentanedionate, Rhodium(III) acetylacetonate

45-1800-250MG 45-1800-1G 45-1800-5G
100196-254EA 401.97 USD
100196-254 100198-202 100207-402
Rhodium (III) acetylacetonate ≥97% (99.9%-Rh)
Rhodium (III) acetylacetonate

Metal Beta-diketonates, Volatile Precursors for CVD

Formula: C₁₅H₂₁O₆Rh
MW: 400.23
Boiling Pt: >280 °C/ 240 °C (0.1 mmHg) (dec./ subl.)
Melting Pt: 260 °C
MDL Number: MFCD00083144
CAS Number: 14284-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR