Rhodium(II) acetate dimer ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Dirhodium tetraacetate, Tetrakis(acetato)dirhodium(II)

45-1730-500MG 45-1730-100MG 45-1730-2G
100206-684EA 1111.08 USD
100206-684 100206-682 100198-024
Rhodium(II) acetate dimer ≥99%
Rhodium (II) acetate dimer
Formula: C₈H₁₂O₈Rh₂
MW: 441.987 g/mol
MDL Number: MFCD00003538
CAS Number: 15956-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR