Rhodium/tetra-n-octylammonium chloride (70-75% Rh), colloidal, purified

Supplier: STREM CHEMICALS INC MS
Warning

45-1660-250MG 45-1660-1G
100196-252EA 859.14 USD
100196-252 100201-230
Rhodium/tetra-n-octylammonium chloride (70-75% Rh), colloidal, purified
Rhodium/tetra-n-octylammonium chloride

Nanomaterials

MDL Number: MFCD06798912
CAS Number: 7440-16-6
UN: 3200
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR