Rhodium (∼9% Rh) (w/w), colloid hydrosol in polyethyleneglycol-dodecylether

Supplier: STREM CHEMICALS INC MS
Warning

45-1550-1G 45-1550-5G
100198-198EA 343.94 USD
100198-198 100198-200
Rhodium (∼9% Rh) (w/w), colloid hydrosol in polyethyleneglycol-dodecylether
Rhodium

Nanomaterials

Formula: Rh
MDL Number: MFCD00011201
CAS Number: 7440-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR