Hydroxy(1,5-cyclooctadiene)rhodium(I) dimer ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

45-1000-250MG 45-1000-1G
100198-808EA 604.37 USD
100198-808 100206-680
Hydroxy(1,5-cyclooctadiene)rhodium(I) dimer ≥97%
Hydroxy(1,5-cyclooctadiene)rhodium(I) dimer
Formula: C₁₆H₂₆O₂Rh₂
MW: 456.1875 g/mol
MDL Number: MFCD04974244
CAS Number: 73468-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR