Carbonylhydridotris(triphenylphosphine)rhodium(I) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tris(triphenylphosphine)rhodium(I) Carbonyl Hydride, Carbonyltris(triphenylphosphine)rhodium(I) Hydride, Hydridocarbonyltris(triphenylphosphine)rhodium(I)

45-0900-5G 45-0900-1G
100203-180EA 1445.11 USD
100203-180 100203-178
Carbonylhydridotris(triphenylphosphine)rhodium(I) ≥98%
Carbonylhydridotris(triphenylphosphine)rhodium(I)
Formula: C₅₅H₄₆OP₃Rh
MW: 918.80 g/mol
Melting Pt: 138 °C (dec.)
MDL Number: MFCD00151644
CAS Number: 17185-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR