Dicarbonyl(2,4-pentanedionato)rhodium(I) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (Acetylacetonato)dicarbonylrhodium(I), Dicarbonylacetylacetonato rhodium(I)

45-0700-250MG 45-0700-5G 45-0700-1G
100209-962EA 308.55 USD
100209-962 100207-688 100209-352
Dicarbonyl(2,4-pentanedionato)rhodium(I) ≥99%
Dicarbonyl(2,4-pentanedionato)rhodium(I)
Formula: C₇H₈O₄Rh
MW: 259.04 g/mol
Melting Pt: 144…147 °C (subl.)
MDL Number: MFCD00009884
CAS Number: 14874-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR