1,5-Cyclooctadiene(hydroquinone)rhodium(I) tetrafluoroborate

Supplier: STREM CHEMICALS INC MS
Warning

45-0652-250MG 45-0652-1G
101419-050EA 135.88 USD
101419-050 101419-052
1,5-Cyclooctadiene(hydroquinone)rhodium(I) tetrafluoroborate
1,5-Cyclooctadiene(hydroquinone)rhodium(I) tetrafluoroborate
MDL Number: MFCD09753039
CAS Number: 120967-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR